Search
Search
| Gene Symbol: PCBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050362.1 | XP_017905851.1 | 2 - 3 | 366 | 96 | |
| XM_018050363.1 | XP_017905852.1 | 3 - 4 | 471 | 96 | ||
| XM_018050364.1 | XP_017905853.1 | 3 - 4 | 424 | 96 | ||
| XM_018050365.1 | XP_017905854.1 | 2 - 3 | 475 | 96 | ||
| XM_018050366.1 | XP_017905855.1 | 2 - 3 | 357 | 96 | ||
| XM_018050367.1 | XP_017905856.1 | 2 - 3 | 472 | 96 | ||
| GenBank mRNA | JO388469.2 | - | 2 - 3 | 311 | 96 | |
| JT714573.1 | - | 2 - 3 | 185 | 96 | ||
Set Membership: |
|