Search
Search
| Gene Symbol: DAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013976750.2 | XP_013832204.1 | 2 - 3 | 470 | 118 | |
| XM_018046258.1 | XP_017901747.1 | 2 - 3 | 478 | 118 | ||
| XM_018046262.1 | XP_017901751.1 | 2 - 3 | 449 | 118 | ||
| XM_018046263.1 | XP_017901752.1 | 2 - 3 | 378 | 118 | ||
| XM_018046264.1 | XP_017901753.1 | 2 - 3 | 367 | 118 | ||
Set Membership: |
|