Search
Search
| Gene Symbol: ABCC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680780.3 | XP_005680837.1 | 23 - 24 | 3469 | 97 | |
| XM_005680781.3 | XP_005680838.1 | 23 - 24 | 3469 | 97 | ||
| XM_005680782.3 | XP_005680839.2 | 23 - 24 | 3468 | 97 | ||
Set Membership: |
|