Search
Search

DYNLL2| Gene Symbol: DYNLL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064111.1 | XP_017919600.1 | 2 - 3 | 418 | 95 | |
| GenBank mRNA | JO431747.2 | - | 2 - 3 | 443 | 95 | |
| JO559349.2 | - | 2 - 3 | 266 | 95 | ||
Set Membership: |
|