Search
Search

BCORL1| Gene Symbol: BCORL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044195.1 | XP_017899684.1 | 13 - 14 | 5232 | 61 | |
| XM_018044196.1 | XP_017899685.1 | 13 - 14 | 5136 | 61 | ||
| XM_018044197.1 | XP_017899686.1 | 12 - 13 | 5513 | 61 | ||
Set Membership: |
|