Search
Search
| Gene Symbol: CERS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047805.1 | XP_017903294.1 | 10 | 1481 | 185 | |
| XM_018047806.1 | XP_017903295.1 | 11 | 1598 | 185 | ||
| XM_018047807.1 | XP_017903296.1 | 10 | 1240 | 185 | ||
| GenBank mRNA | JO644698.2 | - | 11 | 1579 | 185 | |
Set Membership: |
|