Search
Search

BABAM1| Gene Symbol: BABAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013965015.2 | XP_013820469.2 | 2 | 382 | 74 | |
| XM_018051371.1 | XP_017906860.1 | 3 | 488 | 74 | ||
| XM_018051372.1 | XP_017906861.1 | 3 | 504 | 74 | ||
| XM_018051373.1 | XP_017906862.1 | 3 | 467 | 74 | ||
| GenBank mRNA | JO624086.2 | - | 1049 | 74 | ||
Set Membership: |
|