Search
Search

NF1| Gene Symbol: NF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748206.4 | XP_002748252.1 | 29 - 30 | 4356 | 67 | |
| XM_003732962.3 | XP_003733010.1 | 29 - 30 | 4356 | 67 | ||
| XM_008997133.2 | XP_008995381.1 | 30 - 31 | 4331 | 67 | ||
| XM_008997134.2 | XP_008995382.1 | 30 - 31 | 4328 | 67 | ||
| XM_008997135.2 | XP_008995383.1 | 30 - 31 | 5437 | 67 | ||
| XM_017972549.1 | XP_017828038.1 | 29 - 30 | 4356 | 67 | ||
| XM_017972550.1 | XP_017828039.1 | 30 - 31 | 4331 | 67 | ||
| XM_017972551.1 | XP_017828040.1 | 30 - 31 | 4331 | 67 | ||
| XM_017972552.1 | XP_017828041.1 | 29 - 30 | 4356 | 67 | ||
| XM_017972553.1 | XP_017828042.1 | 30 - 31 | 3913 | 67 | ||
Set Membership: |
|