Search
Search

ABCA5| Gene Symbol: ABCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748665.3 | XP_002748711.1 | 15 - 16 | 2151 | 94 | |
| XM_008997648.2 | XP_008995896.1 | 15 - 16 | 2142 | 94 | ||
| XM_017972763.1 | XP_017828252.1 | 16 - 17 | 2222 | 94 | ||
Set Membership: |
|