Search
Search

TBC1D12| Gene Symbol: TBC1D12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756452.4 | XP_002756498.2 | 7 - 8 | 1684 | 88 | |
| XM_017979144.1 | XP_017834633.1 | 6 - 7 | 490 | 88 | ||
Set Membership: |
|