Search
Search

KIT| Gene Symbol: KIT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185038.1 | NP_001171967.1 | 16 - 17 | 2343 | 75 | |
| XM_008993281.2 | XP_008991529.1 | 16 - 17 | 2445 | 75 | ||
| XM_008993282.2 | XP_008991530.1 | 16 - 17 | 2442 | 75 | ||
| XM_008993283.2 | XP_008991531.1 | 16 - 17 | 2442 | 75 | ||
| XM_008993284.2 | XP_008991532.1 | 16 - 17 | 2433 | 75 | ||
| XM_017970039.1 | XP_017825528.1 | 16 - 17 | 2427 | 75 | ||
| XM_017970040.1 | XP_017825529.1 | 16 - 17 | 2506 | 75 | ||
| GenBank mRNA | AB097502.1 | - | 16 - 17 | 2343 | 75 | |
Set Membership: |
|