Search
Search

KIFC3| Gene Symbol: KIFC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008985956.2 | XP_008984204.1 | 4 - 5 | 655 | 66 | |
| XM_017966945.1 | XP_017822434.1 | 6 - 7 | 1069 | 66 | ||
| XM_017966946.1 | XP_017822435.1 | 2 - 3 | 214 | 66 | ||
Set Membership: |
|