Search
Search

SATB1| Gene Symbol: SATB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983880.2 | XP_008982128.1 | 6 - 7 | 1511 | 104 | |
| XM_008983881.2 | XP_008982129.1 | 5 - 6 | 3385 | 104 | ||
| XM_008983882.2 | XP_008982130.1 | 6 - 7 | 1046 | 104 | ||
| XM_008983884.2 | XP_008982132.1 | 6 - 7 | 4403 | 104 | ||
| XM_008983885.2 | XP_008982133.1 | 6 - 7 | 1788 | 104 | ||
| XM_008983886.2 | XP_008982134.1 | 6 - 7 | 3107 | 104 | ||
| XM_008983890.2 | XP_008982138.1 | 5 - 6 | 754 | 104 | ||
| XM_017966009.1 | XP_017821498.1 | 5 - 6 | 3385 | 104 | ||
Set Membership: |
|