Search
Search

DIAPH2| Gene Symbol: DIAPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763068.3 | XP_002763114.1 | 11 - 12 | 1599 | 87 | |
| XM_002763069.4 | XP_002763115.1 | 12 - 13 | 1620 | 87 | ||
| XM_002763070.3 | XP_002763116.1 | 11 - 12 | 1599 | 87 | ||
| XM_008989581.2 | XP_008987829.1 | 12 - 13 | 1620 | 87 | ||
| XM_008989583.2 | XP_008987831.1 | 12 - 13 | 1620 | 87 | ||
| XM_017968317.1 | XP_017823806.1 | 12 - 13 | 1620 | 87 | ||
| XM_017968318.1 | XP_017823807.1 | 11 - 12 | 1599 | 87 | ||
| XM_017968319.1 | XP_017823808.1 | 11 - 12 | 1566 | 87 | ||
| XM_017968320.1 | XP_017823809.1 | 12 - 13 | 1620 | 87 | ||
Set Membership: |
|