Search
Search

IDE| Gene Symbol: IDE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756402.4 | XP_002756448.1 | 9 - 10 | 1707 | 80 | |
| XM_009009970.2 | XP_009008218.1 | 9 - 10 | 1707 | 80 | ||
| XM_009009971.2 | XP_009008219.1 | 9 - 10 | 3025 | 80 | ||
| XM_009009972.2 | XP_009008220.1 | 6 - 7 | 721 | 80 | ||
| XM_017979135.1 | XP_017834624.1 | 9 - 10 | 1707 | 80 | ||
Set Membership: |
|