Search
Search

HSPH1| Gene Symbol: HSPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748952.3 | XP_002748998.1 | 14 - 15 | 2579 | 89 | |
| XM_002748953.3 | XP_002748999.1 | 13 - 14 | 2447 | 89 | ||
Set Membership: |
|