Search
Search

MUSK| Gene Symbol: MUSK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001117.2 | XP_008999365.1 | 12 - 13 | 1566 | 71 | |
| XM_009001134.2 | XP_008999382.1 | 11 - 12 | 1535 | 71 | ||
| XM_009001138.2 | XP_008999386.1 | 11 - 12 | 1532 | 71 | ||
| XM_009001154.2 | XP_008999402.1 | 10 - 11 | 1505 | 71 | ||
| XM_017975456.1 | XP_017830945.1 | 12 - 13 | 1566 | 71 | ||
| XM_017975460.1 | XP_017830949.1 | 11 - 12 | 1536 | 71 | ||
| XM_017975461.1 | XP_017830950.1 | 10 - 11 | 1505 | 71 | ||
Set Membership: |
|