Search
Search

SLC35C2| Gene Symbol: SLC35C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996047.2 | XP_008994295.1 | 7 - 8 | 1013 | 77 | |
| XM_008996048.2 | XP_008994296.1 | 7 - 8 | 1000 | 77 | ||
| XM_008996049.2 | XP_008994297.1 | 7 - 8 | 951 | 77 | ||
| XM_008996050.2 | XP_008994298.1 | 7 - 8 | 940 | 77 | ||
| XM_008996051.2 | XP_008994299.1 | 7 - 8 | 897 | 77 | ||
| XM_008996052.2 | XP_008994300.1 | 7 - 8 | 878 | 77 | ||
| XM_017972080.1 | XP_017827569.1 | 7 - 8 | 1501 | 77 | ||
| XM_017972081.1 | XP_017827570.1 | 5 - 6 | 1027 | 77 | ||
| XM_017972082.1 | XP_017827571.1 | 6 - 7 | 860 | 77 | ||
| XM_017972083.1 | XP_017827572.1 | 7 - 8 | 1233 | 77 | ||
Set Membership: |
|