Search
Search

AGBL5| Gene Symbol: AGBL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017964478.1 | XP_017819967.1 | 5 - 6 | 1767 | 109 | |
| XM_017964479.1 | XP_017819968.1 | 5 - 6 | 1765 | 109 | ||
Set Membership: |
|