Search
Search

SLC41A1| Gene Symbol: SLC41A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760720.3 | XP_002760766.1 | 4 - 5 | 1627 | 72 | |
| XM_008985560.2 | XP_008983808.1 | 11 - 12 | 2182 | 72 | ||
Set Membership: |
|