Search
Search

PSTPIP1| Gene Symbol: PSTPIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008990154.2 | XP_008988402.2 | 4 - 5 | 422 | 70 | |
| XM_017968806.1 | XP_017824295.1 | 4 - 5 | 422 | 70 | ||
| XM_017968807.1 | XP_017824296.1 | 2 - 3 | 254 | 70 | ||
| XM_017968808.1 | XP_017824297.1 | 4 - 5 | 422 | 70 | ||
Set Membership: |
|