Search
Search

ELMOD3| Gene Symbol: ELMOD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017964198.1 | XP_017819687.1 | 8 - 9 | 1407 | 67 | |
| XM_017964199.1 | XP_017819688.1 | 8 - 9 | 953 | 67 | ||
| XM_017964200.1 | XP_017819689.1 | 8 - 9 | 718 | 67 | ||
| XM_017964202.1 | XP_017819691.1 | 7 - 8 | 667 | 67 | ||
| XM_017964203.1 | XP_017819692.1 | 9 - 10 | 1007 | 67 | ||
| XM_017964204.1 | XP_017819693.1 | 6 - 7 | 574 | 67 | ||
| XM_017964205.1 | XP_017819694.1 | 8 - 9 | 718 | 67 | ||
Set Membership: |
|