Search
Search

FNDC3A| Gene Symbol: FNDC3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742665.3 | XP_002742711.2 | 11 - 12 | 2085 | 93 | |
| XM_003731933.3 | XP_003731981.1 | 10 - 11 | 1355 | 93 | ||
Set Membership: |
|