Search
Search

GRM1| Gene Symbol: GRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008995208.1 | XP_008993456.1 | 6 - 7 | 2354 | 63 | |
| XM_008995209.2 | XP_008993457.1 | 6 - 7 | 2357 | 63 | ||
| XM_008995211.1 | XP_008993459.1 | 6 - 7 | 2354 | 63 | ||
Set Membership: |
|