Search
Search

EPB41L2| Gene Symbol: EPB41L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746956.3 | XP_002747002.1 | 8 - 9 | 1396 | 65 | |
| XM_008995050.1 | XP_008993298.1 | 8 - 9 | 1410 | 65 | ||
| XM_008995051.2 | XP_008993299.1 | 8 - 9 | 1859 | 65 | ||
| XM_008995052.1 | XP_008993300.1 | 8 - 9 | 1396 | 65 | ||
| XM_008995054.1 | XP_008993302.1 | 8 - 9 | 1410 | 65 | ||
| XM_008995055.1 | XP_008993303.1 | 8 - 9 | 1410 | 65 | ||
| XM_008995056.1 | XP_008993304.1 | 8 - 9 | 1410 | 65 | ||
| XM_008995057.1 | XP_008993305.1 | 8 - 9 | 1410 | 65 | ||
| XM_008995058.1 | XP_008993306.1 | 8 - 9 | 1410 | 65 | ||
| XM_008995059.1 | XP_008993307.1 | 8 - 9 | 1410 | 65 | ||
| XM_008995060.1 | XP_008993308.1 | 8 - 9 | 1410 | 65 | ||
| XM_008995061.2 | XP_008993309.1 | 8 - 9 | 1672 | 65 | ||
| XM_008995062.1 | XP_008993310.1 | 8 - 9 | 1410 | 65 | ||
| XM_008995063.1 | XP_008993311.1 | 8 - 9 | 1410 | 65 | ||
| XM_008995064.1 | XP_008993312.1 | 8 - 9 | 1410 | 65 | ||
| XM_008995065.1 | XP_008993313.1 | 8 - 9 | 1410 | 65 | ||
| XM_008995067.1 | XP_008993315.1 | 8 - 9 | 1410 | 65 | ||
| XM_008995068.1 | XP_008993316.1 | 8 - 9 | 1396 | 65 | ||
| XM_017971395.1 | XP_017826884.1 | 8 - 9 | 2195 | 65 | ||
Set Membership: |
|