Search
Search

PLCB4| Gene Symbol: PLCB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008995736.2 | XP_008993984.1 | 5 - 6 | 2627 | 69 | |
| XM_008995738.2 | XP_008993986.1 | 8 - 9 | 790 | 69 | ||
| XM_008995739.1 | XP_008993987.1 | 7 - 8 | 638 | 69 | ||
| XM_008995740.2 | XP_008993988.1 | 7 - 8 | 655 | 69 | ||
| XM_008995741.1 | XP_008993989.1 | 6 - 7 | 662 | 69 | ||
| XM_008995742.2 | XP_008993990.1 | 6 - 7 | 3313 | 69 | ||
| XM_008995743.2 | XP_008993991.1 | 4 - 5 | 7603 | 69 | ||
| XM_008995744.2 | XP_008993992.1 | 5 - 6 | 2625 | 69 | ||
| XM_017971954.1 | XP_017827443.1 | 8 - 9 | 710 | 69 | ||
| XM_017971955.1 | XP_017827444.1 | 6 - 7 | 904 | 69 | ||
Set Membership: |
|