Search
Search

SLC4A10| Gene Symbol: SLC4A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749393.2 | XP_002749439.1 | 5 - 6 | 752 | 65 | |
| XM_003733175.2 | XP_003733223.1 | 5 - 6 | 754 | 65 | ||
| XM_008998799.2 | XP_008997047.1 | 5 - 6 | 821 | 65 | ||
| XM_008998800.2 | XP_008997048.1 | 5 - 6 | 822 | 65 | ||
| XM_008998801.2 | XP_008997049.1 | 5 - 6 | 662 | 65 | ||
| XM_008998802.2 | XP_008997050.1 | 5 - 6 | 820 | 65 | ||
| XM_008998803.2 | XP_008997051.1 | 5 - 6 | 821 | 65 | ||
Set Membership: |
|