Search
Search

SLC12A5| Gene Symbol: SLC12A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747609.3 | XP_002747655.1 | 21 - 22 | 2965 | 80 | |
| XM_008996044.1 | XP_008994292.1 | 21 - 22 | 2923 | 80 | ||
Set Membership: |
|