Search
Search

EXOC5| Gene Symbol: EXOC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753942.4 | XP_002753988.1 | 4 - 5 | 722 | 130 | |
| XM_009006081.2 | XP_009004329.1 | 4 - 5 | 589 | 130 | ||
Set Membership: |
|