Search
Search

COG5| Gene Symbol: COG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009002649.2 | XP_009000897.1 | 13 - 14 | 1560 | 117 | |
| XM_009002650.2 | XP_009000898.1 | 13 - 14 | 1560 | 117 | ||
Set Membership: |
|