Search
Search

CLOCK| Gene Symbol: CLOCK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993335.2 | XP_008991583.1 | 8 - 9 | 840 | 103 | |
| XM_008993337.2 | XP_008991585.1 | 8 - 9 | 799 | 103 | ||
| XM_017970557.1 | XP_017826046.1 | 7 - 8 | 609 | 103 | ||
| XM_017970558.1 | XP_017826047.1 | 7 - 8 | 695 | 103 | ||
| XM_017970559.1 | XP_017826048.1 | 8 - 9 | 799 | 103 | ||
Set Membership: |
|