Search
Search

PRPF39| Gene Symbol: PRPF39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753854.3 | XP_002753900.1 | 2 - 3 | 515 | 64 | |
| XM_009005988.2 | XP_009004236.1 | 2 - 3 | 483 | 64 | ||
| XM_009005989.1 | XP_009004237.1 | 2 - 3 | 476 | 64 | ||
Set Membership: |
|