Search
Search

NPAS2| Gene Symbol: NPAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757430.4 | XP_002757476.3 | 5 - 6 | 555 | 88 | |
| XM_017964100.1 | XP_017819589.1 | 5 - 6 | 3992 | 88 | ||
| XM_017964101.1 | XP_017819590.1 | 5 - 6 | 671 | 88 | ||
| XM_017964102.1 | XP_017819591.1 | 2 - 3 | 3609 | 88 | ||
Set Membership: |
|