Search
Search

MYO10| Gene Symbol: MYO10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017969977.1 | XP_017825466.1 | 26 - 27 | 5155 | 61 | |
| XM_017969978.1 | XP_017825467.1 | 26 - 27 | 3864 | 61 | ||
| XM_017969979.1 | XP_017825468.1 | 25 - 26 | 3933 | 61 | ||
Set Membership: |
|