Search
Search

ZDHHC17| Gene Symbol: ZDHHC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752805.3 | XP_002752851.2 | 3 - 4 | 951 | 75 | |
| XM_009004267.2 | XP_009002515.1 | 2 - 3 | 333 | 75 | ||
| XM_017976226.1 | XP_017831715.1 | 3 - 4 | 463 | 75 | ||
Set Membership: |
|