Search
Search

SERTAD4| Gene Symbol: SERTAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760559.3 | XP_002760605.1 | 3 - 4 | 505 | 61 | |
| XM_017966585.1 | XP_017822074.1 | 3 - 4 | 397 | 61 | ||
Set Membership: |
|