Search
Search

SRGAP1| Gene Symbol: SRGAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752697.3 | XP_002752743.1 | 1 - 2 | 240 | 91 | |
| XM_009004152.2 | XP_009002400.1 | 1 - 2 | 240 | 91 | ||
Set Membership: |
|