Search
Search

RGS10| Gene Symbol: RGS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756641.3 | XP_002756687.1 | 2 - 3 | 250 | 106 | |
| XM_009010360.2 | XP_009008608.1 | 2 - 3 | 227 | 106 | ||
Set Membership: |
|