Search
Search

C11H11orf74| Gene Symbol: C11H11orf74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755189.3 | XP_002755235.1 | 5 - 6 | 664 | 115 | |
| XM_009007945.1 | XP_009006193.1 | 5 - 6 | 649 | 115 | ||
| XM_009007946.2 | XP_009006194.1 | 5 - 6 | 914 | 115 | ||
| XM_009007947.2 | XP_009006195.1 | 6 - 7 | 655 | 115 | ||
Set Membership: |
|