Search
Search

RTKN| Gene Symbol: RTKN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757596.3 | XP_002757642.1 | 11 - 12 | 1797 | 82 | |
| XM_008980477.2 | XP_008978725.1 | 12 - 13 | 1907 | 82 | ||
| XM_008980478.2 | XP_008978726.1 | 11 - 12 | 1400 | 82 | ||
| XM_008980479.2 | XP_008978727.1 | 11 - 12 | 1520 | 82 | ||
| XM_008980480.2 | XP_008978728.1 | 10 - 11 | 1587 | 82 | ||
| XM_017964225.1 | XP_017819714.1 | 11 - 12 | 1986 | 82 | ||
| XM_017964226.1 | XP_017819715.1 | 10 - 11 | 1668 | 82 | ||
| GenBank mRNA | EF214855.1 | - | 1 - 2 | 49 | 82 | |
Set Membership: |
|