Search
Search

EMID1| Gene Symbol: EMID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743645.3 | XP_002743691.2 | 12 - 13 | 1257 | 98 | |
| XM_009008714.2 | XP_009006962.1 | 13 - 14 | 1251 | 98 | ||
| XM_009008721.2 | XP_009006969.1 | 13 - 14 | 1239 | 98 | ||
| XM_009008728.2 | XP_009006976.1 | 13 - 14 | 1251 | 98 | ||
| XM_009008733.2 | XP_009006981.1 | 13 - 14 | 1251 | 98 | ||
| XM_009008741.1 | XP_009006989.1 | 12 - 13 | 1257 | 98 | ||
| XM_009008756.2 | XP_009007004.1 | 12 - 13 | 1188 | 98 | ||
| XM_009008763.2 | XP_009007011.1 | 13 - 14 | 1251 | 98 | ||
Set Membership: |
|