Search
Search

SLC20A2| Gene Symbol: SLC20A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757006.4 | XP_002757052.1 | 10 - 11 | 4974 | 90 | |
| XM_008979408.2 | XP_008977656.1 | 10 - 11 | 2235 | 90 | ||
| XM_008979409.2 | XP_008977657.1 | 10 - 11 | 2218 | 90 | ||
| XM_008979410.2 | XP_008977658.1 | 10 - 11 | 2294 | 90 | ||
| XM_008979412.1 | XP_008977660.1 | 10 - 11 | 2190 | 90 | ||
| XM_017963824.1 | XP_017819313.1 | 11 - 12 | 5109 | 90 | ||
Set Membership: |
|