Search
Search

PHF6| Gene Symbol: PHF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763283.4 | XP_002763329.2 | 9 - 10 | 1179 | 102 | |
| XM_008989910.2 | XP_008988158.1 | 9 - 10 | 1182 | 102 | ||
| XM_008989911.2 | XP_008988159.1 | 9 - 10 | 1182 | 102 | ||
| XM_008989913.2 | XP_008988161.1 | 10 - 11 | 1263 | 102 | ||
| XM_017968496.1 | XP_017823985.1 | 9 - 10 | 1182 | 102 | ||
Set Membership: |
|