Search
Search

UST| Gene Symbol: UST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747070.4 | XP_002747116.1 | 6 - 7 | 1252 | 60 | |
| XM_008995219.2 | XP_008993467.1 | 6 - 7 | 1252 | 60 | ||
| XM_008995221.2 | XP_008993469.1 | 6 - 7 | 1252 | 60 | ||
Set Membership: |
|