Search
Search

EMCN| Gene Symbol: EMCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992923.2 | XP_008991171.1 | 8 - 9 | 801 | 83 | |
| XM_008992924.2 | XP_008991172.1 | 8 - 9 | 802 | 83 | ||
| XM_008992925.2 | XP_008991173.1 | 8 - 9 | 803 | 83 | ||
| XM_017970392.1 | XP_017825881.1 | 7 - 8 | 765 | 83 | ||
Set Membership: |
|