Search
Search

PLA2G7| Gene Symbol: PLA2G7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746617.4 | XP_002746663.1 | 4 - 5 | 3128 | 66 | |
| XM_008994532.2 | XP_008992780.1 | 4 - 5 | 698 | 66 | ||
| XM_008994533.2 | XP_008992781.1 | 4 - 5 | 671 | 66 | ||
| XM_017971135.1 | XP_017826624.1 | 5 - 6 | 783 | 66 | ||
| XM_017971136.1 | XP_017826625.1 | 4 - 5 | 3131 | 66 | ||
Set Membership: |
|