Search
Search

FUBP1| Gene Symbol: FUBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750996.3 | XP_002751042.2 | 1 - 2 | 316 | 73 | |
| XM_003733472.3 | XP_003733520.1 | 1 - 2 | 316 | 73 | ||
| XM_009001535.1 | XP_008999783.1 | 1 - 2 | 240 | 73 | ||
| XM_009001536.1 | XP_008999784.1 | 1 - 2 | 240 | 73 | ||
| XM_009001537.1 | XP_008999785.1 | 1 - 2 | 240 | 73 | ||
| XM_009001538.1 | XP_008999786.1 | 1 - 2 | 316 | 73 | ||
| XM_009001540.1 | XP_008999788.1 | 1 - 2 | 316 | 73 | ||
| XM_017974855.1 | XP_017830344.1 | 1 - 2 | 240 | 73 | ||
Set Membership: |
|