Search
Search

SLC37A1| Gene Symbol: SLC37A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761453.3 | XP_002761499.1 | 10 - 11 | 1881 | 69 | |
| XM_008986746.2 | XP_008984994.1 | 10 - 11 | 1645 | 69 | ||
Set Membership: |
|