Search
Search

CASP8AP2| Gene Symbol: CASP8AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994774.1 | XP_008993022.1 | 7 - 8 | 2904 | 80 | |
| XM_008994775.1 | XP_008993023.1 | 7 - 8 | 2863 | 80 | ||
| XM_008994776.1 | XP_008993024.1 | 7 - 8 | 2904 | 80 | ||
| XM_008994777.1 | XP_008993025.1 | 6 - 7 | 2829 | 80 | ||
| XM_008994778.1 | XP_008993026.1 | 6 - 7 | 2762 | 80 | ||
| XM_008994779.1 | XP_008993027.1 | 7 - 8 | 2904 | 80 | ||
Set Membership: |
|