Search
Search

MAPKBP1| Gene Symbol: MAPKBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009005513.2 | XP_009003761.1 | 29 - 30 | 5805 | 53 | |
| XM_009005514.2 | XP_009003762.1 | 31 - 32 | 6456 | 53 | ||
| XM_009005515.2 | XP_009003763.1 | 28 - 29 | 5787 | 53 | ||
| XM_009005516.2 | XP_009003764.1 | 30 - 31 | 6438 | 53 | ||
| XM_017976893.1 | XP_017832382.1 | 30 - 31 | 4909 | 53 | ||
Set Membership: |
|